ICH/USP Oral Target Elements Standard D: Cr @ 11,000; Sn @ 6000; Cu, Mo @ 3000; Ba @ 1400; Sb @ 1200; Li @ 550 µg/mL in 5% HNO3/tr. HF, VHG, LGC Standards
77697-538
New Product
:
- Concentration:Varies
- Matrix:5% HNO₃/tr HF
- Description:ICH/USP Oral Target Elements Standard D: Cr @ 11,000; Sn @ 6000; Cu, Mo @ 3000; Ba @ 1400; Sb @ 1200; Li @ 550 µg/mL in 5% HNO3/tr. HF
- Size:100 ml
- Cat. No.:77697-538