Order Entry
Puerto Rico
ContactUsLinkComponent
ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 µg/mL in 2% HNO3, VHG, LGC Standards
ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-535
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-535
Supplier:  LGC Standards
Supplier Number:  VHG-ICH-USP-TELB-100
New Product
CAS Number:  

Specifications

  • Concentration:
    Varies
  • Matrix:
    2% HNO₃
  • Description:
    ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 µg/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-535

Specifications

About this item