ICH/USP Oral Target Elements Standard AZ: As @ 15; Cd, Pb @ 5 µg/mL in 2% HNO3, VHG, LGC Standards
77697-534
New Product
:
- Concentration:Varies
- Matrix:2% HNO₃
- Description:ICH/USP Oral Target Elements Standard AZ: As @ 15; Cd, Pb @ 5 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77697-534