Order Entry
Puerto Rico
ContactUsLinkComponent
ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 µg/mL in 5% HNO3, VHG, LGC Standards
ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-548
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-548
Supplier:  LGC Standards
Supplier Number:  VHG-ICSAB1-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-548

Specifications

About this item