Order Entry
Puerto Rico
ContactUsLinkComponent
 
Multi-Anion Standard 7A: F-, Cl-, NO3- as N, Br-, SO4(-2), PO4(-3) as P @ 1000 µg/mL in H2O, VHG, LGC Standards
  77697-547
New Product
 :  
undefined
Multi-Anion Standard 7A: F-, Cl-, NO3- as N, Br-, SO4(-2), PO4(-3) as P @ 1000 µg/mL in H2O, VHG, LGC Standards
  77697-547
 :  VHG-ICM7A-100
New Product
 :  

 

  • Concentration:
    1000 µg/mL
  • Matrix:
    H₂O
  • Description:
    Multi-Anion Standard 7A: F-, Cl-, NO3- as N, Br-, SO4(-2), PO4(-3) as P @ 1000 µg/mL in H2O
  • Size:
    100 ml
  • Cat. No.:
    77697-547