ICH/USP Oral Target Elements Standard A: Hg @ 30; As @ 15; Cd, Pb @ 5 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-533
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:2% HNO₃
- Description:ICH/USP Oral Target Elements Standard A: Hg @ 30; As @ 15; Cd, Pb @ 5 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77697-533