Order Entry
Puerto Rico
ContactUsLinkComponent
Phosphate as P @ 10000 µg/mL in H2O, VHG, LGC Standards
Phosphate as P @ 10000 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-532
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Phosphate as P @ 10000 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-532
Supplier:  LGC Standards
Supplier Number:  VHG-I1PPO4P-500
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    H₂O
  • Description:
    Phosphate as P @ 10000 µg/mL in H2O
  • Size:
    500 ml
  • Cat. No.:
    77697-532

Specifications

About this item