Order Entry
Puerto Rico
ContactUsLinkComponent
Rhodium Internal Standard: Rh @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Rhodium Internal Standard: Rh @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Catalog # 77697-633
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Rhodium Internal Standard: Rh @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Catalog # 77697-633
Supplier:  LGC Standards
Supplier Number:  VHG-LISARH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HCl
  • Description:
    Rhodium Internal Standard: Rh @ 10 µg/mL in 2% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-633

Specifications

About this item