Order Entry
Puerto Rico
ContactUsLinkComponent
 
Iridium Internal Standard: Ir @ 10 µg/mL in 2% HCl, VHG, LGC Standards
  77697-632
New Product
 :  
undefined
Iridium Internal Standard: Ir @ 10 µg/mL in 2% HCl, VHG, LGC Standards
  77697-632
 :  VHG-LISAIR-100
New Product
 :  

 

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HCl
  • Description:
    Iridium Internal Standard: Ir @ 10 µg/mL in 2% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-632