Lithium Isotopic Standard: Li @ 100 µg/mL in 2% HNO3 (Enriched in Li-6), VHG, LGC Standards
Catalog # 77697-625
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:2% HNO₃
- Description:Lithium Isotopic Standard: Li @ 100 µg/mL in 2% HNO3 (Enriched in Li-6)
- Size:100 ml
- Cat. No.:77697-625