Order Entry
Puerto Rico
ContactUsLinkComponent
Hafnium Standard: Hf @ 1000 µg/mL in 5% HCl, VHG, LGC Standards
Hafnium Standard: Hf @ 1000 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77697-763
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Hafnium Standard: Hf @ 1000 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77697-763
Supplier:  LGC Standards
Supplier Number:  VHG-PHFH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HCl
  • Description:
    Hafnium Standard: Hf @ 1000 µg/mL in 5% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-763

Specifications

About this item