Order Entry
Puerto Rico
ContactUsLinkComponent
Gallium Standard: Ga @ 1000 µg/mL in 5% HNO3, 0.5% HCl, VHG, LGC Standards
Gallium Standard: Ga @ 1000 µg/mL in 5% HNO3, 0.5% HCl, VHG, LGC Standards
Catalog # 77697-759
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Gallium Standard: Ga @ 1000 µg/mL in 5% HNO3, 0.5% HCl, VHG, LGC Standards
Catalog # 77697-759
Supplier:  LGC Standards
Supplier Number:  VHG-PGANH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃/tr HCl
  • Description:
    Gallium Standard: Ga @ 1000 µg/mL in 5% HNO3, 0.5% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-759

Specifications

About this item