Order Entry
Puerto Rico
ContactUsLinkComponent
Magnesium Standard: Mg @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Magnesium Standard: Mg @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77697-711
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Magnesium Standard: Mg @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77697-711
Supplier:  LGC Standards
Supplier Number:  VHG-OMG-1000-50G
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    oil
  • Description:
    Magnesium Standard: Mg @ 1000 µg/g in Hydrocarbon Oil
  • Size:
    50 ml
  • Cat. No.:
    77697-711

Specifications

About this item