Indium Standard: In @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77697-705
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:5000 µg/mL
- Matrix:oil
- Description:Indium Standard: In @ 5000 µg/g in Hydrocarbon Oil
- Size:50 ml
- Cat. No.:77697-705