Order Entry
Puerto Rico
ContactUsLinkComponent
 
Lubricating Oil Standard 8: Ca @ 0.250; P @ 0.050; S @ 0.125; Zn @ 0.000 wt% in Lubricating Oil, VHG, LGC Standards
  77697-661
New Product
 :  
undefined
Lubricating Oil Standard 8: Ca @ 0.250; P @ 0.050; S @ 0.125; Zn @ 0.000 wt% in Lubricating Oil, VHG, LGC Standards
  77697-661
 :  VHG-LOIL8-100
New Product
 :  

 

  • Concentration:
    Varies
  • Matrix:
    oil
  • Description:
    Lubricating Oil Standard 8: Ca @ 0.250; P @ 0.050; S @ 0.125; Zn @ 0.000 wt% in Lubricating Oil
  • Size:
    100 ml
  • Cat. No.:
    77697-661