Internal Standard Multi-Element Mix 4: In, Tb, Bi @ 10; Ge, Te @ 25; 6Li, Sc @ 50 µg/mL in 5% HNO3, tr. F-, VHG, LGC Standards
Catalog # 77697-623
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:5% HNO₃/tr HF
- Description:Internal Standard Multi-Element Mix 4: In, Tb, Bi @ 10; Ge, Te @ 25; 6Li, Sc @ 50 µg/mL in 5% HNO3, tr. F-
- Size:100 ml
- Cat. No.:77697-623