Order Entry
Puerto Rico
ContactUsLinkComponent
Iridium Standard: Ir @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Iridium Standard: Ir @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Catalog # 77697-619
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Iridium Standard: Ir @ 10 µg/mL in 2% HCl, VHG, LGC Standards
Catalog # 77697-619
Supplier:  LGC Standards
Supplier Number:  VHG-LIRH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HCl
  • Description:
    Iridium Standard: Ir @ 10 µg/mL in 2% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-619

Specifications

About this item