Germanium Standard: Ge @ 10 µg/mL in H2O, tr. F-, VHG, LGC Standards
Catalog # 77697-615
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:H₂O/tr HF
- Description:Germanium Standard: Ge @ 10 µg/mL in H2O, tr. F-
- Size:100 ml
- Cat. No.:77697-615