Order Entry
Puerto Rico
ContactUsLinkComponent
 
Germanium Standard: Ge @ 10 µg/mL in 2% HNO3, tr. HF, VHG, LGC Standards
  77697-614
New Product
 :  
undefined
Germanium Standard: Ge @ 10 µg/mL in 2% HNO3, tr. HF, VHG, LGC Standards
  77697-614
 :  VHG-LGENF-100
New Product
 :  

 

  • Concentration:
    10 µg/mL
  • Description:
    Germanium Standard: Ge @ 10 µg/mL in 2% HNO3, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77697-614