Order Entry
Puerto Rico
ContactUsLinkComponent
Lithium AA Standard: Li @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Lithium AA Standard: Li @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
  77697-461
New Product
 :  
undefined
Lithium AA Standard: Li @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
  77697-461
 :  VHG-AALIN-100
New Product
 :  

 

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Lithium AA Standard: Li @ 1000 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-461