Order Entry
Puerto Rico
ContactUsLinkComponent
Molybdenum AA Standard: Mo @ 1000 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Molybdenum AA Standard: Mo @ 1000 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-465
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Molybdenum AA Standard: Mo @ 1000 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-465
Supplier:  LGC Standards
Supplier Number:  VHG-AAMONF-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃/tr HF
  • Description:
    Molybdenum AA Standard: Mo @ 1000 µg/mL in 5% HNO3, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77697-465

Specifications

About this item