Macitentan, Mikromol, LGC Standards
Catalog # 77697-439
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:441798-33-0
- Description:Macitentan
- Size:250 mg
- Formula:C₁₉H₂₀Br₂N₆O₄S
- Molecular Weight:588.27 g/mol
- Cat. No.:77697-439