Order Entry
Puerto Rico
ContactUsLinkComponent
 
Loteprednol Etabonate, Mikromol, LGC Standards
Catalog # 77697-429
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Loteprednol Etabonate, Mikromol, LGC Standards
Catalog # 77697-429
Supplier:  LGC Standards
Supplier Number:  MM3645.00
New Product
CAS Number:  

Specifications

  • CAS No.:
    82034-46-6
  • Description:
    Loteprednol Etabonate
  • Size:
    250 mg
  • Formula:
    C₂₄H₃₁ClO₇
  • Molecular Weight:
    466.96 g/mol
  • Cat. No.:
    77697-429

Specifications

About this item