$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Methyl 2-(Bromomethyl)-3-nitrobenzoate, Mikromol, LGC Standards
Methyl 2-(Bromomethyl)-3-nitrobenzoate, Mikromol, LGC Standards
77697-428
:
LGC Standards
New Product
:
Methyl 2-(Bromomethyl)-3-nitrobenzoate, Mikromol, LGC Standards
77697-428
:
LGC Standards
:
MM3641.15-0025
New Product
:
CAS No.:
98475-07-1
Description:
Methyl 2-(Bromomethyl)-3-nitrobenzoate
Size:
25 mg
Formula:
C₉H₈BrNO₄
Molecular Weight:
274.07 g/mol
Cat. No.:
77697-428