Lercanidipine Hydrochloride, Mikromol, LGC Standards
Catalog # 77697-418
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:132866-11-6
- Description:Lercanidipine Hydrochloride
- Size:250 mg
- Formula:C₃₆H₄₁N₃O₆·HCl
- Molecular Weight:648.20 g/mol
- Cat. No.:77697-418