Epinastine Hydrochloride, Mikromol, LGC Standards
Catalog # 77697-417
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:108929-04-0
- Description:Epinastine Hydrochloride
- Size:250 mg
- Formula:C₁₆H₁₅N₃
- Molecular Weight:285.78 g/mol
- Cat. No.:77697-417