Methcathinone Hydrochloride (2-Methylamino-1-phenylpropan-1-one Hydrochloride), Mikromol, LGC Standards
Catalog # 77697-406
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:49656-78-2
- Description:Methcathinone Hydrochloride (2-Methylamino-1-phenylpropan-1-one Hydrochloride)
- Size:100 mg
- Formula:C₁₀H₁₄ClNO
- Cat. No.:77697-406