Ibandronate Sodium Monohydrate, Mikromol, LGC Standards
Catalog # 77697-390
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:138926-19-9
- Description:Ibandronate Sodium Monohydrate
- Size:250 mg
- Formula:C₉H₂₂NNaO₇P₂·H₂O
- Molecular Weight:359.23 g/mol
- Cat. No.:77697-390