Hydroxyzine Hydrochloride, Mikromol, LGC Standards
Catalog # 77697-357
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2192-20-3
- Description:Hydroxyzine Hydrochloride
- Size:250 mg
- Formula:C₂₁H₂₇ClN₂O₂·2HCl
- Cat. No.:77697-357