Sodium Fusidate, Mikromol, LGC Standards
Catalog # 77697-354
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:751-94-0
- Description:Sodium Fusidate
- Size:250 mg
- Formula:C₃₁H₄₇NaO₆
- Molecular Weight:538.701 g/mol
- Cat. No.:77697-354