Order Entry
Puerto Rico
ContactUsLinkComponent
Ethyl 8-Hydroxy-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Mikromol, LGC Standards
Ethyl 8-Hydroxy-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Mikromol, LGC Standards
Catalog # 77697-347
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Ethyl 8-Hydroxy-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate, Mikromol, LGC Standards
Catalog # 77697-347
Supplier:  LGC Standards
Supplier Number:  MM0762.02-0025
New Product
CAS Number:  

Specifications

  • CAS No.:
    131666-45-0
  • Description:
    Ethyl 8-Hydroxy-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
  • Size:
    25 mg
  • Formula:
    C₁₅H₁₅N₃O₄
  • Molecular Weight:
    301.29 g/mol
  • Cat. No.:
    77697-347

Specifications

About this item