Pseudooxynicotine Dihydrochloride (4-(Methylamino)-1-(3-pyridyl)butan-1-one Dihydrochloride), Mikromol, LGC Standards
77697-319
New Product
:
- CAS No.:66093-90-1
- Description:Pseudooxynicotine Dihydrochloride (4-(Methylamino)-1-(3-pyridyl)butan-1-one Dihydrochloride)
- Size:25 mg
- Formula:C₁₀H₁₆Cl₂N₂O
- Molecular Weight:251.15 g/mol
- Cat. No.:77697-319