Fexofenadine Hydrochloride, Mikromol, LGC Standards
Catalog # 77697-314
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:153439-40-8
- Description:Fexofenadine Hydrochloride
- Size:250 mg
- Formula:C₃₂H₃₉NO₄
- Molecular Weight:538.13 g/mol
- Cat. No.:77697-314