Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Methyl 4-(Butylamino)benzoate, Mikromol, LGC Standards
Methyl 4-(Butylamino)benzoate, Mikromol, LGC Standards
Methyl 4-(Butylamino)benzoate, Mikromol, LGC Standards
77697-311
:
LGC Standards
New Product
:
Methyl 4-(Butylamino)benzoate, Mikromol, LGC Standards
77697-311
:
LGC Standards
:
MM0504.04
New Product
:
CAS No.:
71839-12-8
Description:
Methyl 4-(Butylamino)benzoate
Size:
100 mg
Formula:
C₁₂H₁₇NO₂
Molecular Weight:
207.269 g/mol
Cat. No.:
77697-311