Indapamide, Mikromol, LGC Standards
Catalog # 77697-302
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:26807-65-8
- Description:Indapamide
- Size:250 mg
- Formula:C₁₆H₁₆ClN₃O₃S
- Molecular Weight:365.83 g/mol
- Cat. No.:77697-302