Nicardipine Hydrochloride, Mikromol, LGC Standards
Catalog # 77697-299
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:54527-84-3
- Description:Nicardipine Hydrochloride
- Size:250 mg
- Formula:C₂₆H₂₉N₃O₆
- Molecular Weight:515.99 g/mol
- Cat. No.:77697-299