Prochlorperazine Maleate, Mikromol, LGC Standards
Catalog # 77697-292
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:84-02-6
- Description:Prochlorperazine Maleate
- Size:250 mg
- Formula:C₂₀H₂₄ClN₃S·2C₄H₄O₄
- Molecular Weight:606.09 g/mol
- Cat. No.:77697-292