$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Ethyl 4-Phenylpiperidine-4-carboxylate, Mikromol, LGC Standards
Ethyl 4-Phenylpiperidine-4-carboxylate, Mikromol, LGC Standards
77697-276
:
LGC Standards
New Product
:
Ethyl 4-Phenylpiperidine-4-carboxylate, Mikromol, LGC Standards
77697-276
:
LGC Standards
:
MM0315.05
New Product
:
CAS No.:
77-17-8
Description:
Ethyl 4-Phenylpiperidine-4-carboxylate
Size:
100 mg
Formula:
C₁₄H₁₉NO₂
Cat. No.:
77697-276