Ethyl 4-(4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate, Mikromol, LGC Standards
77697-264
New Product
:
- CAS No.:165739-83-3
- Description:Ethyl 4-(4,8-Dichloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate
- Size:100 mg
- Formula:C₂₂H₂₂Cl₂N₂O₂
- Molecular Weight:417.33 g/mol
- Cat. No.:77697-264