Fluoxetine Hydrochloride, Mikromol, LGC Standards
Catalog # 77697-260
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:56296-78-7
- Description:Fluoxetine Hydrochloride
- Size:250 mg
- Formula:C₁₇H₁₈F₃NO·HCl
- Molecular Weight:345.79 g/mol
- Cat. No.:77697-260