N-(6-Amino-1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)formamide, Mikromol, LGC Standards
77697-245
New Product
:
- CAS No.:7597-60-6
- Description:N-(6-Amino-1,3-dimethyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)formamide
- Size:100 mg
- Formula:C₇H₁₀N₄O₃
- Molecular Weight:198.18 g/mol
- Cat. No.:77697-245