Nitrendipine, Mikromol, LGC Standards
Catalog # 77697-215
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:39562-70-4
- Description:Nitrendipine
- Size:500 mg
- Formula:C₁₈H₂₀N₂O₆
- Molecular Weight:360.366 g/mol
- Cat. No.:77697-215