Pyridin-2-amine (2-Pyridylamine), Mikromol, LGC Standards
Catalog # 77697-206
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:504-29-0
- Description:Pyridin-2-amine (2-Pyridylamine)
- Size:100 mg
- Formula:C₅H₆N₂
- Molecular Weight:94.12 g/mol
- Cat. No.:77697-206