Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate, Mikromol, LGC Standards
77697-201
New Product
:
- CAS No.:4093-31-6
- Description:Methyl 4-(Acetylamino)-5-chloro-2-methoxybenzoate
- Size:100 mg
- Formula:C₁₁H₁₂ClNO₄
- Molecular Weight:257.67 g/mol
- Cat. No.:77697-201