N,N-Diethylethane-1,2-diamine, Mikromol, LGC Standards
Catalog # 77697-200
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:100-36-7
- Description:N,N-Diethylethane-1,2-diamine
- Size:100 mg
- Formula:C₆H₁₆N₂
- Molecular Weight:116.21 g/mol
- Cat. No.:77697-200