Metamizole Sodium Monohydrate, Mikromol, LGC Standards
Catalog # 77697-194
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:5907-38-0
- Description:Metamizole Sodium Monohydrate
- Size:250 mg
- Formula:C₁₃H₁₆N₃NaO₄S·H₂O
- Molecular Weight:351.36 g/mol
- Cat. No.:77697-194