Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
ent-3-Methoxy-17-methylmorphinan-10-one, Mikromol, LGC Standards
ent-3-Methoxy-17-methylmorphinan-10-one, Mikromol, LGC Standards
ent-3-Methoxy-17-methylmorphinan-10-one, Mikromol, LGC Standards
77697-183
:
LGC Standards
New Product
:
ent-3-Methoxy-17-methylmorphinan-10-one, Mikromol, LGC Standards
77697-183
:
LGC Standards
:
MM0033.02
New Product
:
CAS No.:
57969-05-8
Description:
ent-3-Methoxy-17-methylmorphinan-10-one
Size:
100 mg
Cat. No.:
77697-183