Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
Pyridoxine Hydrochloride, Mikromol, LGC Standards
Pyridoxine Hydrochloride, Mikromol, LGC Standards
77697-178
:
LGC Standards
New Product
:
Pyridoxine Hydrochloride, Mikromol, LGC Standards
77697-178
:
LGC Standards
:
MM0021.00-0250
New Product
:
CAS No.:
58-56-0
Description:
Pyridoxine Hydrochloride
Size:
250 mg
Formula:
C₈H₁₂ClNO₃
Molecular Weight:
205.64 g/mol
Cat. No.:
77697-178