Fluticasone Propionate, LoGiCal, LGC Standards
Catalog # 77697-141
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:80474-14-2
- Description:Fluticasone Propionate
- Size:10 mg
- Formula:C₂₅H₃₁F₃O₅S
- Molecular Weight:500.57 g/mol
- Cat. No.:77697-141