Haloperidol, LoGiCal, LGC Standards
Catalog # 77697-139
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52-86-8
- Description:Haloperidol
- Size:10 mg
- Formula:C₂₁H₂₃ClFNO₂
- Molecular Weight:375.87 g/mol
- Cat. No.:77697-139